Trityl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane
M-Clarity™ quality level = MQ200
T83801-100G T83801-25G T83801-500G
SIALT83801-100GEA 80 EUR
SIALT83801-100G SIALT83801-25G SIALT83801-500G
Trityl chloride, Sigma-Aldrich®
Trityl chloride
Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 109…112 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8,II