Benzophenone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

M-Clarity™ quality level = MQ100

427551-10G 427551-1G
SIAL427551-10GEA 179 EUR
SIAL427551-10G SIAL427551-1G
Benzophenone, Sigma-Aldrich®
Benzophenone
Formula: C₁₃H₁₀O
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR