Benzophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Diphenylmethanone, Diphenyl ketone
M-Clarity™ quality level = MQ300
W213403-10KG-K W213403-1KG-K
SIALW213403-10KG-KEA 464 EUR
SIALW213403-10KG-K SIALW213403-1KG-K
Benzophenone, Sigma-Aldrich®
Benzophenone
Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305.4 °C (1013 hPa)
Melting Pt: 47…49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099