2-Aminobenzophenone, Sigma-Aldrich®

Supplier: Merck

Synonyms: o-Aminobenzophenone

M-Clarity™ quality level = MQ100

A41208-25G A41208-5G
SIALA41208-25GEA 81.5 EUR
SIALA41208-25G SIALA41208-5G
2-Aminobenzophenone, Sigma-Aldrich®
2-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 105…106 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR