4-Aminobenzophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: (4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone
M-Clarity™ quality level = MQ200
A41402-10G A41402-25G
SIALA41402-10GEA 80.7 EUR
SIALA41402-10G SIALA41402-25G
4-Aminobenzophenone, Sigma-Aldrich®
4-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6