2-Hydroxybenzophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone
M-Clarity™ quality level = MQ100
103160-25G 103160-5G
SIAL103160-25GEA 223 EUR
SIAL103160-25G SIAL103160-5G
2-Hydroxybenzophenone, Sigma-Aldrich®
2-Hydroxybenzophenone
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 34…37 °C
Density: 1.03 g/cm³ (172 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7
EINECS: 204-226-2