Dibenzyl ether 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Benzyl ether
148400010 148400025
ACRO148400010EA 48.3 EUR
ACRO148400010 ACRO148400025
Dibenzyl ether 99%
Dibenzyl ether
Formula: C₆H₅CH₂OCH₂C₆H₅
MW: 198.26 g/mol
Boiling Pt: 298
Melting Pt: 1.5…3.5 °C
Density: 1.04 g/cm³
Flash Pt: 135 °C
MDL Number: MFCD00004780
CAS Number: 103-50-4
EINECS: 203-118-2
UN: 3082
ADR: 9 III
Merck Index: 13,01134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb077d69cfe8c3315f62058a54ca8460d752735&vid=08fffe90d8cf2c6ddb596093c7e9631247e2a70b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5610 to 1.5630 (20°C, 589 nm)
GC ≥98.5 %