Triphenylmethane ≥98%
Supplier: Thermo Fisher Scientific
A11710.14 A11710.22 A11710.36
A11710.14EA 29.5 EUR
A11710.14 A11710.22 A11710.36
Triphenylmethane ≥98%
Triphenylmethane
Purity: 98% CAS No.: 519-73-3 Molecular Formula: (C6H5)3CH Formula Weight: 244.34 Melting Point: 92-94° Boiling Point: 358-359° Merck Reference: 14,9741 MDL No.: MFCD00004763
Formula: C₁₉H₁₆
Boiling Pt: 358…359 °C
Melting Pt: 92…94 °C
Density: 1.010
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 14,9741