Triphenylmethane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
101303-100G 101303-25G
SIAL101303-100GEA 86.7 EUR
SIAL101303-100G SIAL101303-25G
Triphenylmethane, Sigma-Aldrich®
Triphenylmethane
Formula: C₁₉H₁₆
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 90…92 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 12,09871