4-Hexylbiphenyl

Supplier: Thermo Scientific

Synonyms: 4-n-Hexylbiphenyl

BTB10235.1 BTB10235.10
MAYBBTB10235.1EA 0 EUR
MAYBBTB10235.1 MAYBBTB10235.10
4-Hexylbiphenyl
4-Hexylbiphenyl
Formula: C₁₈H₂₂
MW: 238.37 g/mol
Boiling Pt: 148
Melting Pt: 352
Density: 0.9875 g/cm³ (20 °C)
MDL Number: MFCD00009527
CAS Number: 59662-31-6
EINECS: 261-845-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR