3,5-Dimethylphenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid

M-Clarity™ quality level = MQ100

480088-25G 480088-5G
SIAL480088-25GEA 258 EUR
SIAL480088-25G SIAL480088-5G
3,5-Dimethylphenylboronic acid, Sigma-Aldrich®
3,5-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR