α-Bromo-m-xylene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methylbenzyl bromide
B83509-25G B83509-5G
SIALB83509-25GEA 52.1 EUR
SIALB83509-25G SIALB83509-5G
α-Bromo-m-xylene, Sigma-Aldrich®
α-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.3711 g/cm³ (23 °C)
MDL Number: MFCD00000177
CAS Number: 620-13-3
EINECS: 210-625-2
UN: 1701
ADR: 6.1,II
Merck Index: 12,10223