3-Bromo-o-xylene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromo-2,3-dimethylbenzene
M-Clarity™ quality level = MQ100
B83789-25G B83789-5G
SIALB83789-25GEA 129 EUR
SIALB83789-25G SIALB83789-5G
3-Bromo-o-xylene, Sigma-Aldrich®
3-Bromo-o-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 214
Density: 1.365 g/cm³ (20 °C)
MDL Number: MFCD00000069
CAS Number: 576-23-8
EINECS: 209-398-2
UN: 1701
ADR: 6.1 II