2-Bromo-p-xylene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene

M-Clarity™ quality level = MQ100

167266-100G 167266-25G
SIAL167266-100GEA 107 EUR
SIAL167266-100G SIAL167266-25G
2-Bromo-p-xylene, Sigma-Aldrich®
2-Bromo-p-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 200
Melting Pt: 8.9
Density: 1.3582 g/cm³ (18.5 °C)
MDL Number: MFCD00000074
CAS Number: 553-94-6
EINECS: 209-055-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR