α'-Bromo-α,α,α-trifluoro-o-xylene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-(Trifluoromethyl)benzyl bromide, 1-(Bromomethyl)-2-(trifluoromethyl)benzene, 2-(Bromomethyl)benzotrifluoride

MQ200

254967-1G 254967-5G
SIAL254967-1GEA 27.5 EUR
SIAL254967-1G SIAL254967-5G
α'-Bromo-α,α,α-trifluoro-o-xylene, Sigma-Aldrich®
α'-Bromo-α,α,α-trifluoro-o-xylene
Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 116…118 °C (60 mmHg)
Melting Pt: 30
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR