1-Bromo-3,5-bis(trifluoromethyl)benzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene
MQ100
290157-10G 290157-50G
SIAL290157-10GEA 54.8 EUR
SIAL290157-10G SIAL290157-50G
1-Bromo-3,5-bis(trifluoromethyl)benzene, Sigma-Aldrich®
1-Bromo-3,5-bis(trifluoromethyl)benzene
Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 152…155 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1.7048 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000381
CAS Number: 328-70-1
EINECS: 206-334-5