4-Bromo-2,6-xylidine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Bromo-2,6-dimethylaniline
M-Clarity™ quality level = MQ200
192376-100G 192376-25G
SIAL192376-100GEA 225 EUR
SIAL192376-100G SIAL192376-25G
4-Bromo-2,6-xylidine, Sigma-Aldrich®
4-Bromo-2,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120 °C (0.2 mmHg)
Melting Pt: 50 °C
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III