α-Chloro-p-xylene, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: alpha-Chloro-p-xylene, 4-Methylbenzyl chloride

M-Clarity™ quality level = MQ100

C73400-100G C73400-5G
SIALC73400-100GEA 56.5 EUR
SIALC73400-100G SIALC73400-5G
α-Chloro-p-xylene, Sigma-Aldrich®
α-Chloro-p-xylene
Formula: C₈H₉Cl
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000919
CAS Number: 104-82-5
EINECS: 203-241-1
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR