2,4-Dichloro-3,5-xylenol

Supplier: Thermo Scientific

Synonyms: 2,4-Dichloro-3,5-dimethylphenol

SB00536.1 SB00536.10 SB00536.50
MAYBSB00536.1EA 0 EUR
MAYBSB00536.1 MAYBSB00536.10 MAYBSB00536.50
2,4-Dichloro-3,5-xylenol
2,4-Dichloro-3,5-xylenol
Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 96 °C
MDL Number: MFCD00019981
CAS Number: 133-53-9
EINECS: 205-109-9
Merck Index: 13,03102

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR