4'-Methylacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Methyl p-tolyl ketone, p-Methylacetophenone, 1-(p-Tolyl)ethanone
M-Clarity™ quality level = MQ300
W267708-5KG-K W267708-10KG-K W267708-1KG-K W267708-25KG-K
SIALW267708-5KG-KEA 485 EUR
SIALW267708-5KG-K SIALW267708-10KG-K SIALW267708-1KG-K SIALW267708-25KG-K
4'-Methylacetophenone, Sigma-Aldrich®
4'-Methylacetophenone
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.013 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8