2,3-Dimethylbenzoyl chloride

Supplier: Thermo Scientific

TL00220.10 TL00220.50
MAYBTL00220.10EA 0 EUR
MAYBTL00220.10 MAYBTL00220.50
2,3-Dimethylbenzoyl chloride
2,3-Dimethylbenzoyl chloride
Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD00045217
CAS Number: 21900-46-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR