2,3-Dimethylbenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

138150-25G 138150-5G
SIAL138150-25GEA 127 EUR
SIAL138150-25G SIAL138150-5G
2,3-Dimethylbenzoic acid, Sigma-Aldrich®
2,3-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 143…147 °C
Density: 1.283 g/cm³ (20 °C)
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR