3'-(Trifluoromethyl)acetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
M-Clarity™ quality level = MQ200
233161-25G 233161-5G
SIAL233161-25GEA 82.3 EUR
SIAL233161-25G SIAL233161-5G
3'-(Trifluoromethyl)acetophenone, Sigma-Aldrich®
3'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198 °C (1013 hPa)
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
EINECS: 206-490-4