4'-(Trifluoromethyl)acetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone
MQ100
233188-25G 233188-5G
SIAL233188-25GEA 164 EUR
SIAL233188-25G SIAL233188-5G
4'-(Trifluoromethyl)acetophenone, Sigma-Aldrich®
4'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 205
Melting Pt: 26…29 °C
Density: 0.924 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0