2,6-Bis(trifluoromethyl)benzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
233196-1G 233196-5G
SIAL233196-1GEA 49.8 EUR
SIAL233196-1G SIAL233196-5G
2,6-Bis(trifluoromethyl)benzoic acid, Sigma-Aldrich®
2,6-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 138…139 °C
MDL Number: MFCD00000376
CAS Number: 24821-22-5
EINECS: 246-479-1