3,5-Bis(trifluoromethyl)benzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
232882-25G 232882-5G
SIAL232882-25GEA 123 EUR
SIAL232882-25G SIAL232882-5G
3,5-Bis(trifluoromethyl)benzoic acid, Sigma-Aldrich®
3,5-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260
Melting Pt: 140…144 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00000388
CAS Number: 725-89-3
EINECS: 211-970-1