α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Supplier: Thermo Scientific
Warning
Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline
SB00812.1 SB00812.50
MAYBSB00812.1EA 0 EUR
MAYBSB00812.1 MAYBSB00812.50
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.4753 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III