α,α,α,α',α',α'-Hexafluoro-3,5-xylidine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline
M-Clarity™ quality level = MQ100
193135-10G 193135-50G
SIAL193135-10GEA 43.5 EUR
SIAL193135-10G SIAL193135-50G
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine, Sigma-Aldrich®
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.4753 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III