2-Iodo-m-xylene
Supplier: Thermo Scientific
DP01104.1 DP01104.10 DP01104.50
MAYBDP01104.1EA 0 EUR
MAYBDP01104.1 MAYBDP01104.10 MAYBDP01104.50
2-Iodo-m-xylene
2-Iodo-m-xylene
Formula: C₈H₉I
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 11.2 °C
Density: 1,6518 g/cm³ (20 °C)
MDL Number: MFCD00019012
CAS Number: 608-28-6