2,6-Diisopropylphenyl isocyanate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,6-Diisopropylphenylisocyanate

M-Clarity™ quality level = MQ100

376930-1G 376930-5G
SIAL376930-1GEA 64.3 EUR
SIAL376930-1G SIAL376930-5G
2,6-Diisopropylphenyl isocyanate, Sigma-Aldrich®
2,6-Diisopropylphenyl isocyanate
Formula: C₁₃H₁₇NO
MW: 203.28 g/mol
Boiling Pt: 117 °C (1013 hPa)
Density: 1.2485 g/cm³ (20 °C)
MDL Number: MFCD00008882
CAS Number: 28178-42-9
EINECS: 248-885-4
UN: 3382
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR