3,5-Xylyl isothiocyanate

Supplier: Thermo Scientific

Synonyms: 3,5-Dimethylphenyl isothiocyanate, 3,5-Dimethylphenylisothiocyanate

TL00501.1 TL00501EA TL00501.50
MAYBTL00501.1EA 0 EUR
MAYBTL00501.1 MAYBTL00501EA MAYBTL00501.50
3,5-Xylyl isothiocyanate
3,5-Xylyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 261 °C (1013 hPa)
MDL Number: MFCD00041082
CAS Number: 40046-30-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR