2-Methylbenzylamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: o-Tolylmethanamine, 2-(Aminomethyl)toluene

127132-1G 127132-5G
SIAL127132-1GEA 25.6 EUR
SIAL127132-1G SIAL127132-5G
2-Methylbenzylamine, Sigma-Aldrich®
2-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 0.9921 g/cm³ (0 °C)
MDL Number: MFCD00008112
CAS Number: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR