4-Methylbenzylamine
Supplier: Thermo Scientific
Danger
Synonyms: 4-(Aminomethyl)toluene
SB01153.1 SB01153.10 SB01153.50
MAYBSB01153.1EA 0 EUR
MAYBSB01153.1 MAYBSB01153.10 MAYBSB01153.50
4-Methylbenzylamine
4-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 195
Melting Pt: 20…21 °C
Density: 0.952 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008123
CAS Number: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8 III