2,6-Diisopropylaniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
157716-100ML 157716-500ML
SIAL157716-100MLEA 48.9 EUR
SIAL157716-100ML SIAL157716-500ML
2,6-Diisopropylaniline, Sigma-Aldrich®
2,6-Diisopropylaniline
Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 257
Melting Pt: –45 °C
Density: 0.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008887
CAS Number: 24544-04-5
EINECS: 246-305-4