2,6-Diisopropylaniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
374733-25G
374733-5G
SIAL374733-25GEA
263
EUR
SIAL374733-25G
SIAL374733-5G
2,6-Diisopropylaniline, Sigma-Aldrich®
2,6-Diisopropylaniline
Formula:
C₁₂H₁₉N Boiling Pt: 257 °C (1013 hPa) Melting Pt: –45 °C Density: 0.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008887 CAS Number: 24544-04-5 EINECS: 246-305-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...