3-Furoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Furan-3-carboxylic acid, 3-Furancarboxylic acid
M-Clarity™ quality level = MQ100
163392-25G
163392-5G
SIAL163392-25GEA
222
EUR
SIAL163392-25G
SIAL163392-5G
3-Furoic acid, Sigma-Aldrich®
3-Furoic acid
Formula:
C₅H₄O₃ |
MDL Number:
MFCD00005350 CAS Number: 488-93-7 EINECS: 207-689-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...