3,4-Xylenol
Supplier: EHRENSTORFER GMBH
EHERC12734000EA
48.4
EUR
EHERC12734000
3,4-Xylenol
3,4-Xylenol
Formula:
C₈H₁₀O MW: 122,17 g/mol Boiling Pt: 227 °C (1013 hPa) Melting Pt: 62…65 °C Density: 1,023 g/cm³ (17 °C) |
MDL Number:
MFCD00002304 UN: 2261 ADR: 6.1,II Merck Index: 12,10215 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...