3,4-Xylenol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIALD175404-500GEA 113 EUR
SIALD175404-500G SIALD175404-5G SIALD175404-100G
3,4-Xylenol, Sigma-Aldrich®
3,4-Xylenol
Formula: C₈H₁₀O
MW: 122,17 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 62…65 °C
Density: 1,023 g/cm³ (17 °C)
MDL Number: MFCD00002304
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR