2,6-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene
M-Clarity™ quality level = MQ100
D175005-100G D175005-500G
SIALD175005-100GEA 28.3 EUR
SIALD175005-100G SIALD175005-500G
2,6-Xylenol, Sigma-Aldrich®
2,6-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 41…42 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002240
CAS Number: 576-26-1
EINECS: 209-400-1
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215