2,6-Xylenol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2,6-Dimethylphenol, 2-Hydroxy-m-xylene, 2-Hydroxy-3-xylene
M-Clarity™ quality level = MQ300
W324906-10KG
W324906-1KG
W324906-5KG
W324906-SAMPLE
SIALW324906-10KGEA
652
EUR
SIALW324906-10KG
SIALW324906-1KG
SIALW324906-5KG
SIALW324906-SAMPLE
2,6-Xylenol, Sigma-Aldrich®
2,6-Xylenol
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 203 Melting Pt: 41…42 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 73 Storage Temperature: Ambient |
MDL Number:
MFCD00002240 CAS Number: 576-26-1 EINECS: 209-400-1 UN: 2261 ADR: 6.1 II Merck Index: 12,10215 |