2,4-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Hydroxy-3-xylene, 2,4-Dimethylphenol, 2,4-Xylenol
M-Clarity™ quality level = MQ100
D174203-100ML D174203-25ML
SIALD174203-100MLEA 115 EUR
SIALD174203-100ML SIALD174203-25ML
2,4-Xylenol, Sigma-Aldrich®
2,4-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 210.9 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.016 g/cm³ (25 °C)
Flash Pt: 80…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002233
CAS Number: 105-67-9
EINECS: 203-321-6
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215