3,5-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 5-Hydroxy-3-xylene, 3,5-Dimethylphenol, 5-Hydroxy-m-xylene, 1,3-Dimethyl-5-hydroxybenzene
M-Clarity™ quality level = MQ100
D175706-100G D175706-500G
SIALD175706-100GEA 53.9 EUR
SIALD175706-100G SIALD175706-500G
3,5-Xylenol, Sigma-Aldrich®
3,5-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 61…64 °C
Density: 0.968 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002307
CAS Number: 108-68-9
EINECS: 203-606-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215