2,5-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,5-Dimethylphenol, 2-Hydroxy-p-xylene, 2-Hydroxy-4-xylene
M-Clarity™ quality level = MQ100
D174602-100G D174602-5G
SIALD174602-100GEA 54.8 EUR
SIALD174602-100G SIALD174602-5G
2,5-Xylenol, Sigma-Aldrich®
2,5-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.169 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002237
CAS Number: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215