Thymol Ph. Eur.

Supplier: VWR Chemicals

83558.180EA 158 EUR
83558.180
Thymol Ph. Eur.
Thymol
Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150,22 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 49…51 °C
Density: 0,965 g/cm³ (25 °C)
Flash Pt: 104 °C
MDL Number: MFCD00002309
UN: 2430
ADR: 8,III
Merck Index: 13,09476
https://si.vwr.com/stibo/web/std.lang.all/78/44/2297844.jpg

Order Now

Specification Test Results

Appearance Colourless crystals
Identification B Passes test
Appearance of solution Passes test
Acidity Passes test
Related substances Passes test
Residue on evaporation Max. 0.05 %
Residual solvents Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR