Thymol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
M-Clarity™ quality level = MQ300
W306606-10KG-K W306606-1KG-K
SIALW306606-10KG-KEA 406 EUR
SIALW306606-10KG-K SIALW306606-1KG-K
Thymol, Sigma-Aldrich®
Thymol
Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 233
Melting Pt: 49…51 °C
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 104
Storage Temperature: Ambient
MDL Number: MFCD00002309
CAS Number: 89-83-8
EINECS: 201-944-8
UN: 2430
ADR: 8 III
Merck Index: 13,09476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3268b34e160525798cab810087412f9b9a9db6&vid=7293cafb537e97291a403e856ca7fdb0223ba714&prid=web&clid=SAPDAM