2,6-Dimethylthiophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3-Dimethyl-2-mercaptobenzene, 2-Mercapto-m-xylene, 2,6-Dimethylbenzene-1-thiol, 2,6-Dimethylbenzenethiol
M-Clarity™ quality level = MQ300
W366609-100G-K W366609-25G-K
SIALW366609-100G-KEA 1270 EUR
SIALW366609-100G-K SIALW366609-25G-K
2,6-Dimethylthiophenol, Sigma-Aldrich®
2,6-Dimethylthiophenol
Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 76 °C (1013 hPa)
Density: 1.038 g/cm³ (25 °C)
MDL Number: MFCD00010021
CAS Number: 118-72-9
EINECS: 204-272-3
UN: 2810
ADR: 6.1,III