o-Ethyltoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Ethyl-2-methylbenzene, 2-Ethyltoluene
M-Clarity™ quality level = MQ100
E49401-25G E49401-5G
SIALE49401-25GEA 205 EUR
SIALE49401-25G SIALE49401-5G
o-Ethyltoluene, Sigma-Aldrich®
o-Ethyltoluene
Formula: C₉H₁₂
Boiling Pt: 164 °C (1013 hPa)
Melting Pt: –17 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009257
CAS Number: 611-14-3
EINECS: 210-255-1
UN: 3295
ADR: 3,III