2,4,6-Trimethylphenylboronic acid

Supplier: Thermo Scientific

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

AC35979.250 AC35979.1
MAYBAC35979.250EA 0 EUR
MAYBAC35979.250 MAYBAC35979.1
2,4,6-Trimethylphenylboronic acid
2,4,6-Trimethylphenylboronic acid
Formula: C₉H₁₃BO₂
MDL Number: MFCD00236060
CAS Number: 5980-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR