2,4,6-Trimethylbenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Mesitaldehyde
M-Clarity™ quality level = MQ100
M6808-10G
M6808-50G
SIALM6808-10GEA
129
EUR
SIALM6808-10G
SIALM6808-50G
2,4,6-Trimethylbenzaldehyde, Sigma-Aldrich®
2,4,6-Trimethylbenzaldehyde
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 14 °C Density: 1.0987 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003341 CAS Number: 487-68-3 EINECS: 207-662-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...