2,4,6-Trimethylaniline, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
132179-100ML
132179-25ML
SIAL132179-100MLEA
52
EUR
SIAL132179-100ML
SIAL132179-25ML
2,4,6-Trimethylaniline, Sigma-Aldrich®
2,4,6-Trimethylaniline
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 233 Melting Pt: –5 °C Density: 0.96 g/cm³ (20 °C) Flash Pt: 96 Storage Temperature: Ambient |
MDL Number:
MFCD00007740 CAS Number: 88-05-1 EINECS: 201-794-3 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...